Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD6 Antibody (96123), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB174388 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB174388 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB174388 Supplier Novus Biologicals Supplier No. FAB727L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD6 Monoclonal antibody specifically detects CD6 in Mouse samples. It is validated for Western Blot, Flow Cytometry

Specifications

Antigen CD6
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 96123
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias CD6 antigenFLJ44171, CD6 molecule, T12, Tp120, TP120T-cell differentiation antigen CD6
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD6, Leu18-Gly396, Accession # Q61003
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 923
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.