Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD6 Antibody (rC6/372), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP320439100UG
Description
CD6 Monoclonal antibody specifically detects CD6 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
CD6 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
CD6 antigenFLJ44171, CD6 molecule, T12, Tp120, TP120T-cell differentiation antigen CD6 | |
Recombinant full-length human CD6 protein | |
100 μg | |
Cytokine Research, Immunology, Signal Transduction | |
923 | |
Store at 4&drg;C. Do not freeze. | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
rC6/372 | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction