Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD6 Antibody (rC6/372), Novus Biologicals™

Mouse Monoclonal Antibody
$216.50 - $454.00
Specifications
Antigen | CD6 |
---|---|
Clone | rC6/372 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
Applications | Immunohistochemistry (Paraffin) |
Description
CD6 Monoclonal antibody specifically detects CD6 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
CD6 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cytokine Research, Immunology, Signal Transduction | |
10mM PBS with 0.05% BSA | |
923 | |
IgG1 κ | |
Protein A or G purified |
rC6/372 | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD6 antigenFLJ44171, CD6 molecule, T12, Tp120, TP120T-cell differentiation antigen CD6 | |
Recombinant full-length human CD6 protein | |
Primary | |
Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title