Learn More
Invitrogen™ CD61 (Integrin beta 3) Monoclonal Antibody (VI-PL2), PerCP-eFluor™ 710, eBioscience™

Description
Description: The VI-PL2 monoclonal antibody reacts with human CD61, also known as integrin beta 3 and GPIIIa. CD61 is a 90-110 kDa member of the beta integrin family expressed by a wide variety of cells, including leukocytes, platelets, endothelial and smooth muscle cells. CD61 binds non-covalently with the alpha integrins CD41 and CD51, to form the alpha IIb beta3 (CD41/CD61) and alpha v beta3 (CD51/CD61) complexes. These alpha beta heterodimers are capable of mediating a variety of cellular responses including adhesion, trafficking, proliferaton and differentiation. The CD41/CD61 integrin has been shown to be involved in platelet aggregation, and binds to fibrinogen, von Willebrand Factor (vWF) and fibronectin. The CD51/CD61 integrin binds to matrix proteins including vitronectin, fibronectin, vWF and fibrinogen, and has been shown to have a strong role in modulating the migration and survival of angiogenic endothelial cells. This VI-PL2 antibody has also been shown to crossreact with canine and several non-human primate species, including baboon, rhesus, and cynomologus monkey. Applications Reported: This VI-PL2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This VI-PL2 antibody has been pre-titrated and tested by flow cytometric analysis of human platelets. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number...
Specifications
Specifications
| Antigen | CD61 (Integrin beta 3) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | VI-PL2 |
| Concentration | 5 μL/Test |
| Conjugate | PerCP-eFluor 710 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | ITGB3 |
| Gene Accession No. | P05106 |
| Gene Alias | antigen CD61; BDPLT16; BDPLT2; beta3 integrin; CD61; EGK_08849; glycoprotein GPIIIa; glycoprotein IIIa; GP3A; GPIIIa; GT; INGRB3; integrin beta 3; integrin beta 3 (Cd61); integrin beta-3; integrin beta-3 subunit; integrin subunit beta 3; integrin, beta 3; integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); ITGB3; platelet glycoprotein IIIa; platelet glycoprotein IIIa integrin subunit beta 3; platelet gpIIIa; platelet membrane glycoprotein IIIa |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.