Learn More
CD61 (Integrin beta 3) Monoclonal Antibody (VI-PL2), Super Bright™ 645, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$402.00
Specifications
Clone | VI-PL2 |
---|---|
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Super Bright 645 |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
64-061-942
![]() |
Invitrogen™
64061942 |
100 Tests |
Each of 1 for $402.00
|
|
|||||
Description
Description: The H1.2F3 monoclonal antibody reacts with mouse CD69, also known as very early activation antigen (VEA). CD69 is approximately 35 kDa and is expressed on the surface as a disulfide-linked dimer. While a small subset of lymphocytes in the thymus, spleen and lymph nodes express this antigen, activation of both T and B cells rapidly upregulates the surface expression of CD69, suggesting a role for CD69 in lymphocyte development and activation. Applications Reported: This H1.2F3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This H1.2F3 antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This may be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 645 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 645 nm. We recommend using a 660/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No.
CD61 (GPIIIa, ITGB3) is a glycoprotein found on megakaryocytes, platelets and their precursors. CD61 antigen plays a role in platelet aggregation and also as a receptor for fibrinogen, fibronectin, von Willebrand factor and vitronectrin. CD61 is a 105 kDa glycoprotein that associates with either the alpha 11b integrin (CD41) or the alpha V integrin (CD51) at the cell surface. CD61 is expressed on platelets and megakaryocytes in association with CD41, and on endothelial cells, monocytes, platelets and osteoclasts in association with CD51. CD61 is a receptor for fibrinogen, fibronectin, Von Willebrand Factor, vitronectin and thrombospondin. The CD61 protein product is composed of an alpha chain and a beta chain. A given chain may combine with multiple partners resulting in different integrins. CD61 is found along with the alpha IIb chain in platelets. Like other integrins, CD61 is known to participate in cell adhesion as well as cell-surface mediated signaling. Diseases associated with CD61 dysfunction include Glanzmann Thrombasthenia and Platelet type-16 Bleeding Disorder.Specifications
VI-PL2 | |
Monoclonal | |
Liquid | |
RUO | |
P05106 | |
ITGB3 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
ITGB3 |
Flow Cytometry | |
Super Bright 645 | |
Mouse | |
Baboon, Canine, Cynomolgus Monkey, Human, Rhesus Macaque | |
102117791, 3690, 403788, 717379 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.