Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD62L Rat anti-Mouse, R-PE, Clone: MEL-14, Southern Biotech™

Rat Monoclonal Antibody
Supplier: Southern Biotechnology Associates 170509
Description
SouthernBiotech offers a variety of anti-mouse primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
| CD62L | |
| Monoclonal | |
| 0.1mg/mL | |
| Supplied in PBS/NaN3 and a stabilizing agent | |
| C3H/eb cloned mouse B cell lymphoma 38C-13 | |
| RUO | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| MEL-14 | |
| R-PE | |
| Rat | |
| 0.1 mg | |
| Primary | |
| Store at 2-8°C; Protect from light; Do not freeze |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction