Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD62L Rat anti-Mouse, R-PE, Clone: MEL-14, Southern Biotech™

Rat Monoclonal Antibody
$288.50 - $382.00
Specifications
Antigen | CD62L |
---|---|
Clone | MEL-14 |
Concentration | 0.1mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
SouthernBiotech offers a variety of anti-mouse primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
CD62L | |
0.1mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
C3H/eb cloned mouse B cell lymphoma 38C-13 | |
Primary |
MEL-14 | |
Flow Cytometry | |
R-PE | |
RUO | |
Supplied in PBS/NaN3 and a stabilizing agent | |
IgG2a | |
Store at 2-8°C; Protect from light; Do not freeze |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title