Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD63 Antibody (2585J), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB296157 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB296157 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB296157 Supplier Novus Biologicals Supplier No. FAB50482N

Rabbit Monoclonal Antibody

CD63 Monoclonal antibody specifically detects CD63 in Human samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Flow (Intracellular), CyTOF

Specifications

Antigen CD63
Applications Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 2585J
Conjugate Alexa Fluor 700
Dilution Western Blot, Simple Western, Immunohistochemistry, Flow (Intracellular), CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD_antigen: CD63, CD63, CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, lysosome-associated membrane glycoprotein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, Tspan30
Host Species Rabbit
Immunogen Chinese Hamster Ovary cell line CHO-derived human CD63, Ala103-Val203, Accession # P08962-1
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cardiovascular Biology, Cellular Signaling, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis
Primary or Secondary Primary
Gene ID (Entrez) 967
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.