Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD63 Antibody (LAMP3/2788), Novus Biologicals™
SDP

Catalog No. NB002964 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002964 20 μg
NB002963 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB002964 Supplier Novus Biologicals Supplier No. NBP27881220UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD63 Monoclonal specifically detects CD63 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.

Specifications

Antigen CD63
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LAMP3/2788
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, Immunohistochemistry 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL
Formulation 10 mM PBS with 0.05% BSA
Gene Alias CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1lysosome-associated membrane glycoprotein 3, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, TSPAN30granulophysin
Gene Symbols CD63
Host Species Mouse
Immunogen Recombinant human CD63 protein fragment (aa100-197) (exact sequence is proprietary)
Quantity 20 μg
Gene ID (Entrez) 967
Test Specificity This CD63 MAb (LAMP3/2788) recognizes protein of 26kDa-60kDa, which is identified as CD63.
Target Species Human, Mouse
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2b κ
Marker Late Endosomes Marker
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.