Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD63 Antibody (LAMP3/529), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N247935APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N247935APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N247935APC7 Supplier Novus Biologicals Supplier No. NBP247935APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD63 Monoclonal specifically detects CD63 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD63
Applications Flow Cytometry
Classification Monoclonal
Clone LAMP3/529
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1lysosome-associated membrane glycoprotein 3, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, TSPAN30granulophysin
Gene Symbols CD63
Host Species Mouse
Immunogen Recombinant human CD63 protein (Uniprot: P08962)
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Autophagy, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis
Primary or Secondary Primary
Gene ID (Entrez) 967.0
Test Specificity This monoclonal antibody recognizes protein of 26kDa-60kDa, which is identified as CD63. Its epitope is different from that of monoclonal antibody LAMP3/803 or LAMP3/968 or NKI/C3 or MX-49.129.5. The tetraspanins are integral membrane proteins expressed on cell surface and granular membranes of hematopoietic cells and are components of multi-molecular complexes with specific integrins. The tetraspanin CD63 is a lysosomal membrane glycoprotein that translocates to the plasma membrane after platelet activation. CD63 is expressed on activated platelets, monocytes and macrophages, and is weakly expressed on granulocytes, T cell and B cells. It is located on the basophilic granule membranes and on the plasma membranes of lymphocytes and granulocytes. CD63 is a member of the TM4 superfamily of leukocyte glycoproteins that includes CD9, CD37 and CD53, which contain four transmembrane regions. CD63 may play a role in phagocytic and intracellular lysosome-phagosome fusion events. CD63 deficiency is associated with Hermansky-Pudlak syndrome and is strongly expressed during the early stages of melanoma progression.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.