Learn More
CD64/IGFR1 Rabbit anti-Human, Polyclonal, Bioss

Description
Specifications
Specifications
| Antigen | CD64/IGFR1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH |
| Gene | FCGR1B |
| Gene Accession No. | P12314, Q92637 |
| Gene Alias | AI323638; AV092959; CD64; CD64 antigen; CD64A; CD64b; Fc fragment of IgG high affinity Ib receptor; Fc fragment of IgG receptor Ia; Fc fragment of IgG receptor Ib; Fc fragment of IgG, high affinity Ia, receptor (CD64); Fc fragment of IgG, high affinity Ia, receptor for (CD64); Fc fragment of IgG, high affinity Ib, receptor (CD64); Fc fragment of IgG, high affinity Ib, receptor for (CD64); Fc gamma receptor; Fc gamma receptor Ia; Fc gamma receptor Ib; Fc receptor, IgG, high affinity I; Fcg1; Fc-gamma receptor 1; Fc-gamma receptor I A1; Fc-gamma receptor I B2; Fc-gamma RI; Fc-gamma RIA; Fc-gamma RIB; FcgammaRI; FcgammaRIa; FCGR1; FCGR1A; FCGR1B; FcgRI; FcRI; FcRIB; FLJ18345; hFcgammaRIB; high affinity IgG Fc receptor, subunit beta; High affinity immunoglobulin gamma Fc receptor I; High affinity immunoglobulin gamma Fc receptor IB; high-affinity immunoglobulin gamma Fc receptor I; IGFR1; IGFRB; IgG Fc receptor I; IgG Fc receptor IB; IgG high affinity Fc receptor; IGGHAFC; RP11-196G18.2 |
| Gene Symbols | FCGR1A, FCGR1B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.