Learn More
CD68 Monoclonal Antibody (FA-11), Alexa Fluor™ 488, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
$187.00 - $421.00
Specifications
| Antigen | CD68 |
|---|---|
| Clone | FA-11 |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
53-068-180
|
Invitrogen™
53068180 |
25 μg |
Each for $187.00
|
|
|||||
53-068-182
![]()
|
Invitrogen™
53068182 |
100 μg |
Each for $421.00
|
|
|||||
Description
This FA-11 monoclonal antibody reacts with mouse CD68, known also as macrosialin. CD68 is a heavily O- and N- glycosylated type I integral membrane protein belonging to the sialomucin family and is closely related to the Lysosomal/endosomal-Associated Mem
CD68 (Macrosialin) is a 110 kDa integral membrane glycoprotein predominantly expressed on the intracellular lysomsomes of monocytes and macrophages and to a lesser extent by dendritic cells and peripheral blood granulocytes. Also, CD68 could play a role in phagocytic activities of tissue macrophages, both in intracellular lysosomal metabolism and extracellular cell-cell and cell-pathogen interactions. CD68 is expressed by interdigitating reticulum cells in tonsil and some histiocytic lymphoma or histiocytosis, acute myeloid leukemia (AML), and granulocytic sarcoma. Elevated expression of CD68 has been demonstrated on CD34+ cells in various human malignancies, including several Acute Myeloid Leukemia studies.Specifications
| CD68 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| CD68; CD68 antigen; CD68 molecule; GP110; Lamp4; macrophage antigen CD68; macrosialin; Scard1; scavenger receptor class D, member 1; similar to CD68 antigen | |
| Cd68 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Cd68 |
| FA-11 | |
| Flow Cytometry | |
| Alexa Fluor 488 | |
| Rat | |
| Mouse | |
| P31996 | |
| 12514 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.