Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD68/SR-D1 Antibody (2449D) [DyLight 488], Novus Biologicals™
SDP

Catalog No. NB402482 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB402482 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB402482 Supplier Novus Biologicals Supplier No. FAB101141K
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD68/SR-D1 Monoclonal antibody specifically detects CD68/SR-D1 in Mouse samples. It is validated for Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen CD68/SR-D1
Applications Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 2449D
Conjugate DyLight 488
Dilution Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD_antigen: CD68, CD68, CD68 antigen, CD68 molecule, DKFZp686M18236, GP110, LAMP4, macrophage antigen CD68, macrosialin, SCARD1, scavenger receptor class D, member 1
Host Species Rabbit
Immunogen Chinese Hamster Ovary cell line, CHO-derived mouse CD68, Asp21-Pro282, Accession # P31996
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytokine Research, Immunology, Microglia Markers
Primary or Secondary Primary
Gene ID (Entrez) 968
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.