Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD68/SR-D1 Antibody (FA-11) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP233337SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP233337SS 0.025 mg
NBP233337 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP233337SS Supplier Novus Biologicals Supplier No. NBP233337SS
Add to Cart
Add to Cart

Rat Monoclonal Antibody has been used in 29 publications

CD68/SR-D1 Monoclonal specifically detects CD68/SR-D1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional, Flow (Intracellular), Single Cell Western.

Specifications

Antigen CD68/SR-D1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone FA-11
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500. Use reported in scientific literature (PMID 34478932), Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (PMID 34478932), Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Functional reported in scientific literature (PMID 11085350), Flow (Intracellular), Single Cell Western
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P31996
Gene Alias CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1
Gene Symbols CD68
Host Species Rat
Immunogen This CD68/SR-D1 Antibody (FA-11) was developed against purified ConA acceptor glycoproteins from the P815 cell line.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cell Biology, Cytokine Research, Immunology, Microglia Markers
Primary or Secondary Primary
Gene ID (Entrez) 968
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.