Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD68/SR-D1 Antibody (FA-11) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody has been used in 29 publications
$268.00 - $572.50
Specifications
Antigen | CD68/SR-D1 |
---|---|
Clone | FA-11 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500. Use reported in scientific literature (PMID 34478932), Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (PMID 34478932), Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Functional reported in scientific literature (PMID 11085350), Flow (Intracellular), Single Cell Western |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP233337SS
![]() |
Novus Biologicals
NBP233337SS |
0.025 mg |
Each for $268.00
|
|
|||||
NBP233337
![]() |
Novus Biologicals
NBP233337 |
0.1 mg |
Each for $572.50
|
|
|||||
Description
CD68/SR-D1 Monoclonal specifically detects CD68/SR-D1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional, Flow (Intracellular), Single Cell Western.Specifications
CD68/SR-D1 | |
1.0 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rat | |
Cell Biology, Cytokine Research, Immunology, Microglia Markers | |
P31996 | |
968 | |
This CD68/SR-D1 Antibody (FA-11) was developed against purified ConA acceptor glycoproteins from the P815 cell line. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
FA-11 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500. Use reported in scientific literature (PMID 34478932), Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (PMID 34478932), Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Functional reported in scientific literature (PMID 11085350), Flow (Intracellular), Single Cell Western | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1 | |
CD68 | |
IgG2a | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title