Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD68/SR-D1 Antibody (OTI1E6), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP270381V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270381V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270381V Supplier Novus Biologicals Supplier No. NBP270381V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD68/SR-D1 Monoclonal specifically detects CD68/SR-D1 in Human samples. It is validated for Western Blot.

Specifications

Antigen CD68/SR-D1
Applications Western Blot
Classification Monoclonal
Clone OTI1E6
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1
Gene Symbols CD68
Host Species Mouse
Immunogen This CD68/SR-D1 Antibody (OTI1E6) was developed against human recombinant protein fragment corresponding to amino acids 22-319 of human CD68 (NP_001242)produced in SF9 cell.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Biology, Cytokine Research, Immunology, Microglia Markers
Primary or Secondary Primary
Gene ID (Entrez) 968.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.