Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD69 Antibody (310106), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB365375 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB365375 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB365375 Supplier Novus Biologicals Supplier No. FAB2386J
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD69 Monoclonal antibody specifically detects CD69 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen CD69
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 310106
Conjugate Janelia Fluor 646
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leukocyte surface antigen Leu-23, MLR-3
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD69, Gly64-Arg199, Accession # P37217
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 969
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.