Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD69 Antibody (FN50), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP143387Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP143387Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP143387Z Supplier Novus Biologicals Supplier No. NBP143387AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD69 Monoclonal specifically detects CD69 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen CD69
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone FN50
Conjugate Alexa Fluor 700
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Gene Alias Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leukocyte surface antigen Leu-23, MLR-3
Gene Symbols CD69
Host Species Mouse
Immunogen The immunogen for this antibody was CD69.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 969
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.