Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD7 Antibody (017), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB074966 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB074966 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB074966 Supplier Novus Biologicals Supplier No. NBP290559DL594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD7 Monoclonal specifically detects CD7 in Mouse samples. It is validated for ELISA.

Specifications

Antigen CD7
Applications ELISA
Classification Monoclonal
Clone 17
Conjugate DyLight 594
Formulation 50 mM Sodium Borate
Gene Alias CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD7 extracellular domain (NP_033984.1; Met 1-Pro 150).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 924
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.