Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD7 Antibody (T3-3A1), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB008475 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB008475 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB008475 Supplier Novus Biologicals Supplier No. NBP254901JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD7 Monoclonal specifically detects CD7 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunofluorescence.

Specifications

Antigen CD7
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone T3-3A1
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein
Gene Symbols CD7
Host Species Mouse
Immunogen Human T cells
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Signal Transduction
Primary or Secondary Primary
Test Specificity Recognizes a protein of 40kDa, identified as CD7, a member of the immunoglobulin gene superfamily. Its N-terminal amino acids 1-107 are highly homologous to Ig kappa-L chains whereas the carboxyl-terminal region of the extracellular domain is proline-rich and has been postulated to form a stalk from which the Ig domain projects. CD7 is expressed on the majority of immature and mature T-lymphocytes, and T cell leukemia. It is also found on natural killer cells, a small subpopulation of normal B cells and on malignant B cells. Cross-linking surface CD7 positively modulates T cell and NK cell activity as measured by calcium fluxes, expression of adhesion molecules, cytokine secretion and proliferation. CD7 associates directly with phosphoinositol 3'-kinase. CD7 ligation induces production of D-3 phosphoinositides and tyrosine phosphorylation.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.