Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD7 Antibody (V55P2F2*B12), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP250142AI Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250142AI 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250142AI Supplier Novus Biologicals Supplier No. NBP250142AF700

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

CD7 Monoclonal specifically detects CD7 in Fish samples. It is validated for ELISA.

Specifications

Antigen CD7
Applications ELISA
Classification Monoclonal
Clone V55P2F2*B12
Conjugate Alexa Fluor 700
Dilution ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein
Gene Symbols CD7
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 924
Target Species Fish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.