Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD72.1 Rat anti-Mouse, Clone: 4G10, Novus Biologicals™
SDP

Catalog No. NBP23604005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP23604005 0.05 mg
NB019658 0.02 mg
NBP236040 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP23604005 Supplier Novus Biologicals Supplier No. NBP2360400.05MG

Rat Monoclonal Antibody

CD72.1 Monoclonal specifically detects CD72.1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen CD72.1
Applications Western Blot
Classification Monoclonal
Clone 4G10
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000
Formulation PBS with no preservative
Gene Alias CD72 antigen, Ly19, Ly32, Lyb2, Lym19
Gene Symbols Cd72
Host Species Rat
Immunogen Mouse recombinant protein CD72
Quantity 0.05 mg
Primary or Secondary Primary
Gene ID (Entrez) 12517
Reconstitution Reconstitute with 0.2 mL sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.