Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD72 Antibody (BU40), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. N106430A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N106430A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N106430A70 Supplier Novus Biologicals Supplier No. NB10064350AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD72 Monoclonal specifically detects CD72 in Human, Rhesus Macaque samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot.

Specifications

Antigen CD72
Applications CyTOF, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone BU40
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD72 antigenB-cell differentiation antigen CD72, CD72 molecule, CD72b, Lyb-2, LYB2lyb-2
Gene Symbols CD72
Host Species Mouse
Immunogen HF B1 plasmacytoid cell line. Considered to be late B like.
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 971
Target Species Human, Rhesus Macaque
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.