Learn More
CD72 Mouse anti-Human, Unconjugated, Clone: 2C1C6, Proteintech
Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 674241IG150UL
Description
The CD72 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD72. This antibody recognizes human antigen. The CD72 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
CD72 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA | |
2C1C6 | |
Unconjugated | |
150 μL | |
971 | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.