Learn More
CD72 Mouse anti-Human, Unconjugated, Clone: 2C1C6, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | CD72 |
|---|---|
| Clone | 2C1C6 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-9526
![]()
|
Proteintech Group Inc
674241IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The CD72 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD72. This antibody recognizes human antigen. The CD72 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
| CD72 | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 971 | |
| Primary |
| 2C1C6 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA | |
| Unconjugated | |
| Human | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.