Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD74 Antibody (3D7), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP259697G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259697G 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259697G Supplier Novus Biologicals Supplier No. NBP259697G

Mouse Monoclonal Antibody

CD74 Monoclonal specifically detects CD74 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen CD74
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 3D7
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD74 antigen, CD74 antigen (invariant polypeptide of major histocompatibility complex, classII antigen-associated), CD74 molecule, major histocompatibility complex, class II invariant chain, DHLAGgamma chain of class II antigens, HLA class II histocompatibility antigen gamma chain, HLADG, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia antigen-associated invariant chain, Ia-associated invariant chain, Ia-GAMMA, Ii, MHC HLA-DR gamma chain, p33
Gene Symbols CD74
Host Species Mouse
Immunogen Human CD74 invariant chain synthetic peptide.
Purification Method Protein G purified
Quantity 100 μL
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 972
Test Specificity Detects approx 80, 55kDa. No cross-reactivity against Cav Beta4.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.