Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD74 Antibody (CLIP/3127R), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB389326 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB389326 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB389326 Supplier Novus Biologicals Supplier No. NBP308927AF700
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD74 Monoclonal antibody specifically detects CD74 in Human, Mouse, Baboon, Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)

Specifications

Antigen CD74
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CLIP/3127R
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias CD74 antigen, CD74 antigen (invariant polypeptide of major histocompatibility complex, classII antigen-associated), CD74 molecule, major histocompatibility complex, class II invariant chain, DHLAGgamma chain of class II antigens, HLA class II histocompatibility antigen gamma chain, HLADG, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia antigen-associated invariant chain, Ia-associated invariant chain, Ia-GAMMA, Ii, MHC HLA-DR gamma chain, p33
Host Species Rabbit
Immunogen SU-DHL-4 lymphoma cells
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 972
Target Species Human, Mouse, Baboon, Rat (Negative)
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.