Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD74 Mouse anti-Human, Clone: 2D8A1, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 663901IG150UL
Description
Expression of the invariant chain of the HLA-DR antigen (CD74) by B-lymphocytes begins in the pre-B stage but lost before the final differentiation to plasma cells. This protein is expressed in B-lymphocytes and many B-cell lymphomas.Specifications
| CD74 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P04233 | |
| CD74 | |
| CD74 Fusion Protein Ag25503 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 2D8A1 | |
| Unconjugated | |
| CD74 | |
| CD74, DHLAG, HLADG, Ia GAMMA, Ii, p33 | |
| Mouse | |
| Protein A | |
| RUO | |
| 972 | |
| -20°C | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction