Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD74 Mouse anti-Human, Clone: 2D8A1, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | CD74 |
|---|---|
| Clone | 2D8A1 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6819
![]()
|
Proteintech Group Inc
663901IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Expression of the invariant chain of the HLA-DR antigen (CD74) by B-lymphocytes begins in the pre-B stage but lost before the final differentiation to plasma cells. This protein is expressed in B-lymphocytes and many B-cell lymphomas.Specifications
| CD74 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CD74, DHLAG, HLADG, Ia GAMMA, Ii, p33 | |
| CD74 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 2D8A1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P04233 | |
| 972 | |
| CD74 Fusion Protein Ag25503 | |
| Primary | |
| -20°C | |
| CD74 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title