Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD79a Monoclonal Antibody (HM57)
GREENER_CHOICE

Catalog No. p-7171262
Change view
Click to view available options
Quantity:
100 μg
200 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA181870 100 μg
PIMA516710 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA181870 Supplier Invitrogen™ Supplier No. MA181870
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Membrane permeabilization is required for flow cytometry applications. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. A suggested positive control for immunohistochemical applications is human tonsil. This product requires antigen retrieval using heat treatment prior to staining of paraffin sections. Mouse anti Human CD79a antibody, clone HM57 recognizes an epitope within the cytoplasmic domain of CD79a.

CD79a is a disulphide-linked heterodimer that includes B29 (CD79b) polypeptide. CD79a is a B lymphocyte antigen receptor with an antigen-specific surface component Ig (immunoglobulin) that associates with Ig-alpha and Ig-beta, necessary elements for the expression and function of the B-cell antigen receptor. CD79a first appears at pre-B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines. Diseases associated with CD79a include Agammaglobulinemia 3 and Agammaglobulinemia, Non-Bruton type.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD79a
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone HM57
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CD79A
Gene Accession No. P11911, P11912
Gene Alias B-cell antigen receptor; B-cell antigen receptor complex-associated protein alpha chain; B-cell antigen receptor complex-associated protein alpha chain-like; CD79; CD79a; CD79A antigen (immunoglobulin-associated alpha); CD79a molecule; CD79a molecule, immunoglobulin-associated alpha; CD79A protein; CD79-alpha protein; EGK_10665; Ig alpha; IGA; Igalpha; ig-alpha; immunoglobulin-associated alpha; LOC100913063; Ly54; Ly-54; MB1; MB-1; MB-1 membrane glycoprotein; Membrane-bound immunoglobulin-associated protein; surface IgM-associated protein
Gene Symbols CD79A
Host Species Mouse
Immunogen Synthetic peptide corresponding to 202-216 amino acid sequence of human mb-1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100913063, 12518, 973
Target Species Human, Mouse, Rat, Multi-species
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.