Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD79B Antibody (071), DyLight 350
SDP

Catalog No. NB076734 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB076734 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB076734 Supplier Novus Biologicals Supplier No. NBP290706UV

Rabbit Monoclonal Antibody

CD79B Monoclonal specifically detects CD79B in Mouse samples. It is validated for ELISA.

Specifications

Antigen CD79B
Applications ELISA
Classification Monoclonal
Clone 71
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD79B (P15530; Met1-Asp158).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 974
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.