Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD79B Antibody (HM79-11), PerCP, Novus Biologicals™
SDP

Catalog No. NB10065611P Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065611P 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065611P Supplier Novus Biologicals Supplier No. NB10065611PCP
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

CD79B Monoclonal specifically detects CD79B in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.

Specifications

Antigen CD79B
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone HM79-11
Conjugate PerCP
Dilution Flow Cytometry
Gene Alias B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein
Gene Symbols CD79B
Host Species Armenian Hamster
Immunogen CD79 alpha/CD79 beta heterodimer purified from WEH1231 B cells.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 974
Test Specificity NB100-65611 recognizes murine CD79 beta, which is expressed by B cells as part of the B cell receptor complex (immunoglobulin and the CD79 alpha/beta heterodimer). Murine CD79 beta is expressed at the cell surface of pro-B cells prior to surface immunoglobulin, and is expressed throughout B cell differentiation. CD79 beta is a B cell specific marker, valuable for the detection of B cells at all maturation stages.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.