Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD79B Antibody (rIGB/1842), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB387433 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387433 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387433 Supplier Novus Biologicals Supplier No. NBP308357R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD79B Monoclonal antibody specifically detects CD79B in Human samples. It is validated for ELISA

Specifications

Antigen CD79B
Applications ELISA
Classification Monoclonal
Clone rIGB/1842
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein
Host Species Mouse
Immunogen Recombinant human CD79B protein fragment (around aa29-159) (exact sequence is proprietary) (Uniprot: P40259)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 974
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.