Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Antibody (12.C7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB100640210.025MG
Description
CD8 Monoclonal specifically detects CD8 in Human, Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CD8 | |
Monoclonal | |
1.0 mg/mL | |
Flow Cytometry 1:100-1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
CD8, CD8a molecule, LEU2, p32 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Recognizes the rabbit CD8 cell surface antigen, expressed by a subset of T lymphocytes with cytotoxic/suppressor activity. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry, Immunocytochemistry | |
12.C7 | |
Unconjugated | |
Phosphate buffered saline with 0.09% Sodium Azide | |
CD8A | |
Made to Rabbit CD8 | |
0.025 mg | |
Innate Immunity, Stem Cell Markers | |
925 | |
Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction