Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Antibody (12.C7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
$206.50 - $487.50
Specifications
Antigen | CD8 |
---|---|
Clone | 12.C7 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry 1:100-1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017557
![]() |
Novus Biologicals
NB100640210.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10064021
![]() |
Novus Biologicals
NB10064021 |
0.1 mg |
Each for $487.50
|
|
|||||
Description
CD8 Monoclonal specifically detects CD8 in Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
CD8 | |
1.0 mg/mL | |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Innate Immunity, Stem Cell Markers | |
925 | |
Made to Rabbit CD8 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
12.C7 | |
Flow Cytometry 1:100-1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
CD8, CD8a molecule, LEU2, p32 | |
CD8A | |
IgG1 | |
Protein G purified | |
Recognizes the rabbit CD8 cell surface antigen, expressed by a subset of T lymphocytes with cytotoxic/suppressor activity. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title