Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Antibody (14), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP250467G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250467G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250467G Supplier Novus Biologicals Supplier No. NBP250467G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD8 Monoclonal specifically detects CD8 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.

Specifications

Antigen CD8
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry
Classification Monoclonal
Clone 14
Conjugate DyLight 488
Dilution Flow Cytometry, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD8, CD8a molecule, LEU2, p32
Gene Symbols CD8A
Host Species Mouse
Immunogen Fibronectin-purified human monocytes
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cytokine Research, Immunology, Immunophenotyping, Innate Immunity, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 925
Test Specificity suitability for western blotting not known
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.