Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Antibody (2ST8.5H7), Novus Biologicals™
SDP

Catalog No. NB017757 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017757 0.025 mg
NB10065928 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017757 Supplier Novus Biologicals Supplier No. NB100659280.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD8 Monoclonal specifically detects CD8 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Western Blot.

Specifications

Antigen CD8
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2ST8.5H7
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry Neat-1:10, Immunoprecipitation 1:10-1:500
Formulation PBS and 1% BSA. with No Preservative
Gene Accession No. P10966
Gene Alias CD8, CD8a molecule, LEU2, p32
Gene Symbols CD8A
Host Species Mouse
Immunogen Normal human T lymphocytes
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cytokine Research, Immunology, Immunophenotyping, Innate Immunity, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 925
Test Specificity NB100-65928 recognizes the human CD8 cell surface antigen. The antibody binds to the beta chain, recognizing an epitope that is dependent upon expression of both CD8 alpha and CD8 beta. CD8 molecules are composed of dimers of either alpha/alpha or alpha/beta polypeptide combinations. Both types of dimer may co-exist at the cell surface.
Target Species Human, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.