Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Antibody (37006R) [DyLight 550], Novus Biologicals™
SDP

Catalog No. NB399798 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB399798 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB399798 Supplier Novus Biologicals Supplier No. FAB1509RL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD8 Monoclonal antibody specifically detects CD8 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen CD8
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 37006R
Conjugate DyLight 550
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD_antigen: CD8a, CD8, CD8 antigen, alpha polypeptide (p32), CD8A, CD8a molecule, LEU2, Leu2 T-lymphocyte antigen, MAL, OKT8 T-cell antigen, p32, T cell co-receptor, T8 T-cell antigen, T-cell antigen Leu2, T-cell surface glycoprotein CD8 alpha chain, T-lymphocyte differentiation antigen T8/Leu-2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD8 alpha , Ser22-Val198, Accession # NP_741969
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 925
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.