Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Antibody (4B11), Novus Biologicals™
SDP

Catalog No. NB10065729 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065729 0.1 mL
NB165729SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10065729 Supplier Novus Biologicals Supplier No. NB10065729
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 4 publications

CD8 Monoclonal specifically detects CD8 in Human samples. It is validated for Dual RNAScope ISH-IHC, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).

Specifications

Antigen CD8
Applications Dual RNAScope ISH-IHC, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4B11
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:20-1:40, Immunohistochemistry-Frozen 1:20-1:40, Dual RNAscope ISH-IHC
Formulation Tissue culture supernatant with 0.09% Sodium Azide
Gene Accession No. P01732
Gene Alias CD8, CD8a molecule, LEU2, p32
Gene Symbols CD8A
Host Species Mouse
Immunogen Synthetic peptide derived from the carboxy terminal region of the humanCD8 alpha chain coupled to a N-terminal cysteine, with the sequence C-KSDGKPSLSARYV. The peptide was coupled to bovine serum albuman and keyhole limpet hemocyanin.
Molecular Weight of Antigen 26 kDa
Purification Method Tissue culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 925
Test Specificity Recognizes the human CD8 cell surface antigen, a 32kD glycoprotein expressed by the cytotoxic/suppressor subset of T-cells and weakly by NK cells. Clone 4B11 has been reported as being suitable for use in Western blotting.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.