Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Antibody (4B11), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 4 publications
$206.50 - $501.50
Specifications
| Antigen | CD8 |
|---|---|
| Clone | 4B11 |
| Dilution | Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:20-1:40, Immunohistochemistry-Frozen 1:20-1:40, Dual RNAscope ISH-IHC |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In Situ Hybridization (ISH) |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB165729SS
![]() |
Novus Biologicals
NB10065729SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB10065729
![]() |
Novus Biologicals
NB10065729 |
0.1 mL |
Each for $501.50
|
|
|||||
Description
CD8 Monoclonal specifically detects CD8 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dual RNAscope ISH-IHC.Specifications
| CD8 | |
| Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:20-1:40, Immunohistochemistry-Frozen 1:20-1:40, Dual RNAscope ISH-IHC | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Human | |
| P01732 | |
| 925 | |
| Synthetic peptide derived from the carboxy terminal region of the humanCD8 alpha chain coupled to a N-terminal cysteine, with the sequence C-KSDGKPSLSARYV. The peptide was coupled to bovine serum albuman and keyhole limpet hemocyanin. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 26 kDa |
| 4B11 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In Situ Hybridization (ISH) | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers | |
| Tissue culture supernatant with 0.09% Sodium Azide | |
| CD8, CD8a molecule, LEU2, p32 | |
| CD8A | |
| IgG2b | |
| Tissue culture supernatant | |
| Recognizes the human CD8 cell surface antigen, a 32kD glycoprotein expressed by the cytotoxic/suppressor subset of T-cells and weakly by NK cells. Clone 4B11 has been reported as being suitable for use in Western blotting. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title