Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Antibody (53-6.7) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP149045SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149045SS 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149045SS Supplier Novus Biologicals Supplier No. NBP149045SS
Add to Cart
Add to Cart

Rat Monoclonal Antibody has been used in 16 publications

CD8 Monoclonal specifically detects CD8 in Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Cell depletion, Inhibition of T Cell Function, CyTOF-ready.

Specifications

Antigen CD8
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 53-6.7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature, Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 24565643), Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Cell depletion, Inhibition of T Cell Function, CyTOF-ready
Formulation PBS with 0.02% Sodium Azide
Gene Alias 53-6.7, 53-6.7 CD8, 53-6.7 CD8 Alpha, 53-6.7 CD8 Aplha, 53-6.7 Clone, CD8, CD8a molecule, Cytotoxic T cell marker, Cytotoxic T lymphocyte marker, LEU2, p32, T cell marker
Gene Symbols CD8A
Host Species Rat
Immunogen Mouse thymus or spleen.
Molecular Weight of Antigen 27 kDa
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 925
Target Species Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.