Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Antibody (53-6.7), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB174929 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB174929 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB174929 Supplier Novus Biologicals Supplier No. FAB116D
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD8 Monoclonal specifically detects CD8 in Mouse samples. It is validated for Cell depletion, CyTOF, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Inhibition of T Cell Function.

Specifications

Antigen CD8
Applications Cell Depletion, CyTOF, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Inhibition Assays
Classification Monoclonal
Clone 53-6.7
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias CD_antigen: CD8a, CD8, CD8 antigen, alpha polypeptide (p32), CD8A, CD8a molecule, LEU2, Leu2 T-lymphocyte antigen, MAL, OKT8 T-cell antigen, p32, T cell co-receptor, T8 T-cell antigen, T-cell antigen Leu2, T-cell surface glycoprotein CD8 alpha chain, T-lymphocyte differentiation antigen T8/Leu-2
Host Species Rat
Immunogen Mouse thymus or spleen
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 925
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.