Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Antibody (C8/144B), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NBP234588J Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234588J 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP234588J Supplier Novus Biologicals Supplier No. NBP234588APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD8 Monoclonal specifically detects CD8 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD8
Applications Flow Cytometry
Classification Monoclonal
Clone C8/144B
Conjugate APC
Dilution Flow Cytometry
Gene Alias CD8, CD8a molecule, LEU2, p32
Gene Symbols CD8A
Host Species Mouse
Immunogen A 13 amino acid peptide from C-terminal cytoplasmic domain of alpha chain of human CD8 molecule. (Uniprot: P01732)
Molecular Weight of Antigen 32 kDa
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 925
Test Specificity CD8 is a cell surface receptor expressed either as a heterodimer with the CD8 beta chain (CD8 alpha/beta) or as a homodimer (CD8 alpha/alpha). A majority of thymocytes and a subpopulation of mature T cells and NK cells express CD8a. CD8 binds to MHC class 1 and through its association with protein tyrosine kinase p56lck plays a role in T cell development and activation of mature T cells. For mature T-cells, CD4 and CD8 are mutually exclusive, so anti-CD8, generally used in conjunction with anti-CD4. It is a useful marker for distinguishing helper/inducer T-lymphocytes, and most peripheral T-cell lymphomas are CD4+/CD8-. Anaplastic large cell lymphoma is usually CD4+ and CD8-, and in T-lymphoblastic lymphoma/leukemia, CD4 and CD8 are often co-expressed. CD8 is also found in littoral cell angioma of the spleen.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.