Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Antibody (EP72), FITC, Novus Biologicals™
SDP

Catalog No. NBP128336 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128336 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128336 Supplier Novus Biologicals Supplier No. NBP128336

Mouse Monoclonal Antibody

CD8 Monoclonal specifically detects CD8 in Chicken samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.

Specifications

Antigen CD8
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone EP72
Concentration 0.5mg/mL
Conjugate FITC
Dilution Flow Cytometry <= 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Gene Alias CD8, CD8a molecule, LEU2, p32
Gene Symbols CD8A
Host Species Mouse
Immunogen Chicken splenocytes
Purification Method Protein A or G purified
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Innate Immunity, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 925
Test Specificity alpha chain of chicken CD8 (Mr 34 kDa)
Target Species Chicken
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.