Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Antibody (RPA-T8) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP225195 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225195 0.1 mg
NBP225195SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP225195 Supplier Novus Biologicals Supplier No. NBP225195
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 7 publications

CD8 Monoclonal specifically detects CD8 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, In vitro assay, Flow (Cell Surface), CyTOF-ready.

Specifications

Antigen CD8
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), In vitro Assay, CyTOF
Classification Monoclonal
Clone RPA-T8
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.1-0.25 ug per 1x10^6 cells, Immunohistochemistry, Immunohistochemistry-Frozen reported in scientific literature (PMID 7882470), In vitro assay reported in multiple pieces of scientific literature, Flow (Cell Surface), CyTOF-ready
Gene Accession No. NP_741969
Gene Alias CD8, CD8 alpha flow, CD8a molecule, LEU2, p32, RPA-T8 RPA-T8 antibody flow, RPA-T8 CD8, RPA-T8 Clone
Gene Symbols CD8A
Host Species Mouse
Immunogen CD8 (CD8a; alpha chain)
Molecular Weight of Antigen 26 kDa
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 925
Test Specificity This product reacts with Human.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.