Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Antibody (YTC182.20), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N064922P594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N064922P594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N064922P594 Supplier Novus Biologicals Supplier No. NB10064922PEATT594
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD8 Monoclonal specifically detects CD8 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen CD8
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone YTC182.20
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD8, CD8a molecule, LEU2, p32
Gene Symbols CD8A
Host Species Rat
Immunogen Mouse L cells transfected with human CD8 gene
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Cytokine Research, Immunology, Immunophenotyping, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 925.0
Test Specificity Recognizes the human CD8 cell surface antigen, a 30/32kD glycoprotein expressed by the cytotoxic/suppressor subset of T lymphocytes, and more weakly by NK cells.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.