Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 beta Antibody (039), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB109146 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB109146 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB109146 Supplier Novus Biologicals Supplier No. NBP289751DL594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD8 beta Monoclonal specifically detects CD8 beta in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CD8 beta
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 39
Conjugate DyLight 594
Formulation 50 mM Sodium Borate
Gene Alias CD8 antigen, beta polypeptide 1 (p37), CD8b antigen, CD8b molecule, CD8B1P37, LEU2, LY3, LYT3, MGC119115, T lymphocyte surface glycoprotein beta chain, T-cell surface glycoprotein CD8 beta chain
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CD8 beta (P10966-1; Met1-Pro170).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 926
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.