Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 beta Antibody (CBB/1595), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB381426 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB381426 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB381426 Supplier Novus Biologicals Supplier No. NBP308335JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD8 beta Monoclonal antibody specifically detects CD8 beta in Bovine, Goat, Sheep samples. It is validated for ELISA

Specifications

Antigen CD8 beta
Applications ELISA
Classification Monoclonal
Clone CBB/1595
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias CD8 antigen, beta polypeptide 1 (p37), CD8b antigen, CD8b molecule, CD8B1P37, LEU2, LY3, LYT3, MGC119115, T lymphocyte surface glycoprotein beta chain, T-cell surface glycoprotein CD8 beta chain
Host Species Mouse
Immunogen Antibody was raised against bovine CD8b. (Uniprot: P10966)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 926
Target Species Bovine, Goat, Sheep
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.