Learn More
Description
Specifications
Specifications
| Antigen | CD8 beta |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | EP42 |
| Conjugate | APC |
| Dilution | Flow Cytometry |
| Gene Alias | CD8 antigen, beta polypeptide 1 (p37), CD8b antigen, CD8b molecule, CD8B1P37, LEU2, LY3, LYT3, MGC119115, T lymphocyte surface glycoprotein beta chain, T-cell surface glycoprotein CD8 beta chain |
| Gene Symbols | CD8B |
| Host Species | Mouse |
| Immunogen | Chicken splenocytes |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
