Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 beta Antibody (EP42), PerCP, Novus Biologicals™
SDP

Catalog No. NBP128300P Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128300P 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128300P Supplier Novus Biologicals Supplier No. NBP128300PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD8 beta Monoclonal specifically detects CD8 beta in Chicken samples. It is validated for Flow Cytometry.

Specifications

Antigen CD8 beta
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone EP42
Conjugate PerCP
Dilution Flow Cytometry
Gene Alias CD8 antigen, beta polypeptide 1 (p37), CD8b antigen, CD8b molecule, CD8B1P37, LEU2, LY3, LYT3, MGC119115, T lymphocyte surface glycoprotein beta chain, T-cell surface glycoprotein CD8 beta chain
Gene Symbols CD8B
Host Species Mouse
Immunogen Chicken splenocytes
Purification Method Protein A or G purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 926
Test Specificity beta chain (Mr 34 kDa) of chicken CD8
Target Species Chicken
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.